missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, ReagentPlus™, Honeywell™
Reagent Grade, 99%
$527.28 - $527.28
Chemical Identifiers
| CAS | 7681-11-0 |
|---|---|
| Molecular Formula | IK |
| Molecular Weight (g/mol) | 166.003 |
| MDL Number | MFCD00011405 |
| InChI Key | NLKNQRATVPKPDG-UHFFFAOYSA-M |
| Synonym | potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide |
| PubChem CID | 4875 |
| ChEBI | CHEBI:8346 |
| IUPAC Name | potassium;iodide |
| SMILES | [K+].[I-] |
Description
- ≥95% for general laboratory use.
Chemical Identifiers
| 7681-11-0 | |
| 166.003 | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| 4875 | |
| potassium;iodide |
| IK | |
| MFCD00011405 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| CHEBI:8346 | |
| [K+].[I-] |
Specifications
| 7681-11-0 | |
| IK | |
| MFCD00011405 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| potassium;iodide | |
| 4875 | |
| 166g/mol | |
| ReagentPlus | |
| Potassium Iodide |
| 500 g | |
| KI | |
| NONH for all modes of transport | |
| Soluble in water | |
| [K+].[I-] | |
| 166.003 | |
| CHEBI:8346 | |
| 99% | |
| 1mmHg (745°C) |
Safety and Handling
P305 + P351 + P338
H302-H315-H319
EINECSNumber : 231-659-4
RTECSNumber : TT2975000