missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™

Catalog No. 60045815
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
6 x 100 g
6 x 250 g
6 x 500 g
6 x 1 kg
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60045815 6 x 500 g
6002256 100 g
6002258 250 g
6002259 500 g
60045812 6 x 100 g
6002257 1 kg
60045814 6 x 250 g
6002260 5 kg
60045813 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 60045815 Supplier Honeywell Chemicals Supplier No. 303156X500G
Only null left

Puriss. p.a., Reag. ISO, Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1,323°C
Melting Point 681°C (literature)
CAS Max % 100.0000%
Quantity 6 x 500 g
Linear Formula KI
Formula Weight 166
Percent Purity >99.5%
Grade puriss. p.a.
Packaging Poly Bottle
Density 3.12 g/cm3
Chemical Name or Material Potassium iodide, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥99.5%
Show More Show Less
ShelfLife 1260 days from date of manufacture
EINECSNumber 231-659-4
Recommended Storage Room Temp