missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium iodide, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™
Puriss. p.a., Reag. ISO, Reag. Ph. Eur.,≥99.5%
Supplier: Honeywell Chemicals 303156X1KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| 1,323°C | |
| 681°C (literature) | |
| 7681-11-0 | |
| 6 x 1 kg | |
| KI | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| [K+].[I-] | |
| 166.003 | |
| CHEBI:8346 | |
| >99.5% | |
| Poly Bottle |
| Potassium iodide | |
| Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥99.5% | |
| 100.0000% | |
| IK | |
| MFCD00011405 | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| potassium;iodide | |
| 4875 | |
| 166 | |
| puriss. p.a. | |
| 3.12 g/cm3 |
Chemical Identifiers
| 7681-11-0 | |
| 166.003 | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| 4875 | |
| potassium;iodide |
| IK | |
| MFCD00011405 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| CHEBI:8346 | |
| [K+].[I-] |
Safety and Handling
ShelfLife : 1260 days from date of manufacture
EINECSNumber : 231-659-4
Recommended Storage : Room Temp