missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, Puratronic™, 99.998% (metals basis)

Catalog No. AA1084206
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1084206 5 g
AA1084222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1084206 Supplier Thermo Scientific Chemicals Supplier No. 01084206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7681-11-0 | IK | 166.003 g/mol

It is both a source of iodide and a mild reducing agent. It is used as a component in the electrolyte of dye sensitized solar cells. In the field of synthetic organic chemistry, it is used in the preparation of alkyl or aryl iodo compounds. It is employed as a nucleophilic catalyst in the alkylations with alkyl bromide, chloride, tosylate and mesylate. In the presence of acid it forms hydroiodic acid, which is a strong reducing agent. It is useful in producing Lugol’s solution (or Lugol’s iodne) which finds many applications including starch testing, gram stains, and etching of gold surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1330°C
Melting Point 681°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.998% (metals basis)
Solubility Information Soluble in water,and acetone. Slightly soluble in ether,and ammonia.
Formula Weight 166.01
Odor Odorless
Refractive Index 1.677
Sensitivity Hygroscopic
Appearance White crystalline powder
Vapor Pressure 1.33 hpa (1mm Hg) at 745°C
Density 3.12
Chemical Name or Material Potassium iodide
Show More Show Less
Hazard Category H372
Precautionary Statement P260-P264b-P270-P314-P501c
EINECSNumber 213-659-4
RTECSNumber TT2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only