missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide (Granular, Free Flowing/Certified ACS), Fisher Chemical™

Catalog No. p-2431398
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
10 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P410100 100 g Poly Bottle
P410500 500 g Poly Bottle
P4103 3 kg Poly Bottle
P41010 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. P410100 Supplier Thermo Fisher Scientific Supplier No. FLP410100
Only null left

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1330°C
Melting Point 680°C
Color White
pH 6 to 8
Physical Form Solid
Quantity 100 g
Assay Percent Range ≥99 %
Formula Weight 166
Percent Purity ≥99%
Grade Certified ACS
Identification Pass Test
Loss on Drying 0.2% max.
Packaging Poly Bottle
Vapor Pressure 1mmHg at 745°C
Density 3.13g/mL
Chemical Name or Material Potassium Iodide
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:1
Instability:0