missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, Crystal, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P133525KGGL

 View more versions of this product

Catalog No. 18612780


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7681-11-0
6.0 to 9.2
IK
[K+].[I-]
166.00
ACS
Amber Glass Bottle
1
2.5 kg
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide
99%
0.002

Chemical Identifiers

7681-11-0
166.00
potassium iodide
IK
NLKNQRATVPKPDG-UHFFFAOYSA-M
[K+].[I-]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.