missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, Crystal, ACS, 99%, Spectrum™ Chemical

Catalog No. 18612777
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612777 25 g Amber Glass Bottle
18612778 125 g Amber Glass Bottle
18612779 500 g Amber Glass Bottle
18612780 2.5 kg Amber Glass Bottle
18612781 2.5 kg Poly Bottle
18612782 12 kg Poly Pail
18612783 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18612777 Supplier Spectrum Chemical Mfg Cor Supplier No. P133525GM
Only null left

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.00
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
IUPAC Name potassium iodide
SMILES [K+].[I-]

Specifications

CAS Max % 1
pH 6.0 to 9.2
Quantity 25 g
Percent Purity 99%
Grade ACS
Loss on Drying 0.002
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.