missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide concentrate, Honeywell™

Catalog No. 6002667
Change view
Click to view available options
Quantity:
1 Pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002667 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6002667 Supplier Honeywell Chemicals Supplier No. 381101EA
Only null left

for 1L standard solution, 0.1 M KI

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Quantity 1 Pc.
Linear Formula KI
UN Number NONH for all modes of transport
Formula Weight 166g/mol
Chemical Name or Material Potassium iodide concentrate