missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, Certified, 5.0% (w/v) ±0.1%, LabChem™
Supplier: LabChem LC197404
Specifications
| Potassium Iodide | |
| Yellow | |
| Liquid | |
| IK | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| potassium;iodide | |
| 4875 | |
| 166 | |
| Certified | |
| Poly Bottle | |
| Iodine vapor |
| 7681-11-0,7732-18-5 | |
| 95,5 | |
| 4 L | |
| KI | |
| Soluble in water | |
| [K+].[I-] | |
| 166.003 | |
| CHEBI:8346 | |
| 5.0% ±0.1% (w/v) | |
| Passes Test | |
| 1.04g/mL | |
| 1.04g/mL |
Chemical Identifiers
| 7681-11-0 | |
| 166.003 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| CHEBI:8346 | |
| [K+].[I-] |
| IK | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| 4875 | |
| potassium;iodide |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature; Protect from light