missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, ACS Reagent Grade, Ricca Chemical
Specifications
Specifications
| Color | White |
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Physical Form | Crystals or Powder |
| Quantity | 100 g |
| Percent Purity | ≥ 99% |
| Grade | ACS |
| Packaging | White Poly Wide Mouth Bottle |
| Chemical Name or Material | Potassium Iodide |