missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, ACS Grade, ≥ 99.0%, LabChem™
Specifications
Specifications
| Boiling Point | 1,330°C |
| Melting Point | 680°C |
| Color | White |
| CAS Max % | 100 |
| Physical Form | Crystals |
| Quantity | 100 g |
| Linear Formula | KI |
| Solubility Information | Soluble in water |
| Formula Weight | 166 |
| Percent Purity | ≥99.0% |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes eye irritation. |
| Precautionary Statement | GHS P Statement Wash exposed skin thoroughly after handling. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
| EINECSNumber | 231-659-4 |
| Recommended Storage | Room Temperature; Protect from light |