missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, 99%, extra pure, briquettes

Catalog No. p-4509784
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC196731000 100 g Glass bottle
AC196735000 500 g Glass bottle
AC196730025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC196731000 Supplier Thermo Scientific Chemicals Supplier No. 196731000
Only null left

CAS: 7681-11-0 | IK | 166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1330.0°C
Melting Point 680.0°C
Color White
Physical Form Briquettes
Quantity 100 g
Assay Percent Range 98.5% min. (on dried substance) (Argentometry)
Linear Formula KI
Fieser 06,227; 11,44
Merck Index 15,7764
Solubility Information Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol
Formula Weight 166
Percent Purity 99%
Grade Extra Pure
Loss on Drying 1% max.
Packaging Glass bottle
Chemical Name or Material Potassium iodide
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 231-659-4

RUO – Research Use Only