missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Potassium iodide, 99%, for biochemistry
CAS: 7681-11-0 | IK | 166 g/mol
$216.80 - $216.80
Chemical Identifiers
| CAS | 7681-11-0 |
|---|---|
| Molecular Formula | IK |
| Molecular Weight (g/mol) | 166 |
| InChI Key | NLKNQRATVPKPDG-UHFFFAOYSA-M |
| Synonym | potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide |
| PubChem CID | 4875 |
| ChEBI | CHEBI:8346 |
| IUPAC Name | potassium;iodide |
| SMILES | [K+].[I-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC447111000
|
Thermo Scientific Chemicals
447111000 |
100 g |
Each for $216.80
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7681-11-0 | |
| 166 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| CHEBI:8346 | |
| [K+].[I-] |
| IK | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| 4875 | |
| potassium;iodide |
Specifications
| 1330.0°C | |
| 7681-11-0 | |
| 98.5% min. (Argentometry) | |
| KI | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| potassium;iodide | |
| 4875 | |
| 166 | |
| Biochemical | |
| Potassium iodide |
| 680.0°C | |
| 100 g | |
| IK | |
| 15,7764 | |
| Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol | |
| [K+].[I-] | |
| 166 | |
| CHEBI:8346 | |
| 99% | |
| Glass bottle |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 231-659-4
RUO – Research Use Only