missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium iodide, 99+%, ACS reagent
CAS: 7681-11-0 | IK | 166 g/mol
Supplier: Thermo Scientific Chemicals 418265000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1330.0°C | |
| 680.0°C | |
| 7681-11-0 | |
| 6.0 to 9.2 (5% soln. at 25°C) | |
| 500 g | |
| IK | |
| MFCD00011405 | |
| 15, 7764 | |
| Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol | |
| [K+].[I-] | |
| 166.003 | |
| CHEBI:8346 | |
| ≥99% | |
| 0.2% max. (150°C, 6 hrs) | |
| 3.13 |
| Potassium iodide | |
| granular | |
| White | |
| Crystals or Granules | |
| 99+% | |
| KI | |
| 06,227; 11,440 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| potassium;iodide | |
| 4875 | |
| 166 | |
| ACS Reagent | |
| 1mmHg at 745°C |
Chemical Identifiers
| 7681-11-0 | |
| MFCD00011405 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| CHEBI:8346 | |
| [K+].[I-] |
| IK | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| 4875 | |
| potassium;iodide |
Safety and Handling
MOISTURE SENSITIVE
LIGHT SENSITIVE
EINECSNumber : 231-659-4
RTECSNumber : TT2975000
TSCA : TSCA
RUO – Research Use Only