missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, 99.995%, (trace metal basis), extra pure

Catalog No. p-3734787
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC193790100 10 g Glass bottle
AC193790500 50 g Glass bottle
AC193792500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC193790100 Supplier Thermo Scientific Chemicals Supplier No. 193790100
Only null left

CAS: 7681-11-0 | IK | 166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1330.0°C
Melting Point 680.0°C
Color White
Physical Form Crystals or Powder
Quantity 10 g
Assay Percent Range 99.995%
Linear Formula KI
Fieser 06,227; 11,440
Merck Index 15, 7764
Solubility Information Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol
Formula Weight 166
Percent Purity 99.995%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Potassium iodide
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 231-659-4

RUO – Research Use Only