missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, 99.0% min., ACS, MilliporeSigma™

$1980.33 - $9997.31

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MPX15055
View Documents
Technidata
PX15055
2.5 kg
Each for $1,980.33
Only null left
 
MPX15057
View Documents
Technidata
PX15057
12 kg
Each for $9,997.31
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7681-11-0
166.003
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
CHEBI:8346
[K+].[I-]
IK
NLKNQRATVPKPDG-UHFFFAOYSA-M
4875
potassium;iodide

Specifications

White
Crystals or Granules
99.0% min.
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
[K+].[I-]
166.003
CHEBI:8346
Potassium Iodide, For ACS analysis
7681-11-0
2.5 kg
IK
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium;iodide
4875
166
Videos
SDS
Documents

Documents