missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Chloride Buffer Solution, pH 10.0, EP, Ricca Chemical
Spécifications
Spécifications
| Chemical Name or Material | Ammonium Chloride Buffer Solution |
| Color | Colorless |
| CAS | 7732-18-5, 1336-21-6, 12125-02-9 |
| Boiling Point | Approximately 100°C |
| Melting Point | ∼0°C |
| Grade | Laboratory |
| Odor | Colorless |
| Packaging | Natural Poly Bottle |
| pH | 10 |
| Quantity | 500 mL |
| Afficher plus de résultats |