missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodate-Iodide, Certified, 0.1000N ±0.0005N (0.0167M), LabChem™
Supplier: LabChem LC196802
Specifications
| Potassium Iodate-Iodide | |
| 96.04,0.36,3.5,0.1 | |
| Liquid | |
| KIO3 | |
| Soluble in water | |
| Certified | |
| Passes Test | |
| Iodine vapor; Carbon dioxide; Carbon monoxide |
| Colorless | |
| 7758-05-6,7681-11-0,1310-73-2 | |
| 1 L | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
| 214 | |
| 0.1000N ±0.0005N (0.0167M) | |
| Amber Poly |