missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate-Iodide, Certified, 0.1000N ±0.0005N (0.0167M), LabChem™

Catalog No. p-5388178
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC196801 500 mL
LC196802 1 L
LC196804 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC196801 Supplier LabChem Supplier No. LC196801
Only null left

Specifications

Chemical Name or Material Potassium Iodate-Iodide
Color Colorless
CAS 7758-05-6, 7681-11-0, 1310-73-2
CAS Max % 96.04, 0.36, 3.5, 0.1
Physical Form Liquid
Quantity 500 mL
Molecular Formula IKO3
Linear Formula KIO3
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
Solubility Information Soluble in water
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
SMILES [O-]I(=O)=O.[K+]
IUPAC Name potassium;iodate
Molecular Weight (g/mol) 214.00
PubChem CID 23665710
Formula Weight 214
Grade Certified
Concentration or Composition (by Analyte or Components) 0.1000N ±0.0005N (0.0167M)
Identification Passes Test
Packaging Amber Poly
Decomposition Information Iodine vapor; Carbon dioxide; Carbon monoxide
Show More Show Less