missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate-Iodide, (1mL = 0.5mg SO32-), for Sulfite, Certified, 0.0125N ±0.0002N (0.002083M) , LabChem™

Supplier:  LabChem LC196602

 View more versions of this product

Catalog No. LC196602


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Iodate-Iodide
(1mL = 0.5mg SO32-), for Sulfite
7681-11-0,7758-05-6,144-55-8
0.0125N ±0.0002N (0.002083M)
Soluble in water
Certified
Amber Poly
Iodine vapor; Carbon dioxide; Carbon monoxide
Liquid
Colorless to Yellow
99.49,0.44,0.04,0.03
KIO3
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
214
Passes Test
Traceable to NIST
1 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions