missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodate-Iodide, (1mL = 0.5mg SO32-), for Sulfite, Certified, 0.0125N ±0.0002N (0.002083M) , LabChem™
Supplier: LabChem LC196601
Specifications
| Potassium Iodate-Iodide | |
| (1mL = 0.5mg SO32-), for Sulfite | |
| 7681-11-0,7758-05-6,144-55-8 | |
| 0.0125N ±0.0002N (0.002083M) | |
| Soluble in water | |
| Certified | |
| Amber Poly | |
| Iodine vapor; Carbon dioxide; Carbon monoxide | |
| Liquid |
| Colorless to Yellow | |
| 99.49,0.44,0.04,0.03 | |
| KIO3 | |
| water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
| 214 | |
| Passes Test | |
| Traceable to NIST | |
| 500 mL |