missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate-Iodide, 0.0125 N (N/80), 1 mL = 0.5 mg SO32-, Ricca Chemical

Catalog No. R62800004F
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R62800004F 4 L Cubitainer
628016 500 mL Amber Glass Bottle
628032 1 L Amber Glass Bottle
62801 4 L Amber Glass Bottle
62805 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. R62800004F Supplier Ricca Chemical Company Supplier No. R62800004F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Potassium Iodate-Iodide
Color Colorless
CAS 7758-05-6, 7681-11-0, 7758-05-6, 1310-73-2
CAS Min % 98.0, 0.42, 0.05, 0.0016
CAS Max % 100.0, 0.44, 0.05, 0.0016
Molecular Formula IKO3
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
SMILES [O-]I(=O)=O.[K+]
IUPAC Name potassium;iodate
Molecular Weight (g/mol) 214.00
PubChem CID 23665710
Equivalence 1 mL = 0.5 mg SO
Grade Laboratory
Packaging Cubitainer
pH Slightly Alkaline
Quantity 4 L
Physical Form Liquid
Concentration or Composition Notes 0.0125Normal
Show More Show Less