missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium iodate concentrate, For 1L standard solution, 1/60Â M KIO3 (0.1N), Honeywell Fluka™
for 1L standard solution, 1/60Â M KIO3 (0.1N)
$206.90 - $206.90
Chemical Identifiers
| Molecular Formula | IKO3 |
|---|---|
| Molecular Weight (g/mol) | 214.00 |
| MDL Number | MFCD00011406 |
| InChI Key | JLKDVMWYMMLWTI-UHFFFAOYSA-M |
| Synonym | potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat |
| PubChem CID | 23665710 |
| IUPAC Name | potassium;iodate |
| SMILES | [O-]I(=O)=O.[K+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| IKO3 | |
| MFCD00011406 | |
| potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
| potassium;iodate |
| 214.00 | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| 23665710 | |
| [O-]I(=O)=O.[K+] |
Specifications
| 5-6-7758 | |
| IKO3 | |
| MFCD00011406 | |
| potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
| [O-]I(=O)=O.[K+] | |
| 214.00 | |
| 214g/mol | |
| Potassium iodate concentrate |
| 1 Pc. | |
| KIO3 | |
| NONH for all modes of transport | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| potassium;iodate | |
| 23665710 | |
| Ampoule |