missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodate concentrate, For 1L standard solution, 1/60 M KIO3 (0.1N), Solstice

Catalog No. 6002668
Change view
Click to view available options
Quantity:
1 Pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002668 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6002668 Supplier Solstice Supplier No. 381201EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

for 1L standard solution, 1/60 M KIO3 (0.1N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
MDL Number MFCD00011406
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]

Specifications

CAS 5-6-7758
Quantity 1 Pc.
Linear Formula KIO3
UN Number NONH for all modes of transport
Formula Weight 214g/mol
Packaging Ampoule
Chemical Name or Material Potassium iodate concentrate