missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodate, Certified by BAM, ≥99.5%, MilliporeSigma™ Supelco™

Catalog No. 111018351
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018351 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018351 Supplier Merck Emd Millipore Supplier No. 6038650GF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 5-6-7758
Density 3.93 g/mL (at 25°C)
Physical Form Neat
Linear Formula KIO3
MDL Number MFCD00011406
Melting Point 560°C
Type Certified Reference Material
Quantity 50 g
Formula Weight 214.00
Percent Purity ≥99.5%
Grade Certified Reference Material
Show More Show Less