missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodate, Certified, 0.1000N ±0.0005N (0.0167M), LabChem™
Supplier: LabChem LC196302
Specifications
| Potassium Iodate | |
| Colorless | |
| Liquid | |
| IKO3 | |
| potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| potassium;iodate | |
| 23665710 | |
| Certified | |
| Passes Test | |
| 1g/mL | |
| 1g/mL |
| 7758-05-6,7732-18-5 | |
| 99.64,0.36 | |
| 1 L | |
| KIO3 | |
| Soluble in water | |
| [O-]I(=O)=O.[K+] | |
| 214.00 | |
| 214 | |
| 0.1000N ±0.0005N (0.0167M) | |
| Poly Bottle | |
| Iodine vapor |
Chemical Identifiers
| 7758-05-6 | |
| 214.00 | |
| potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
| potassium;iodate |
| IKO3 | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| 23665710 | |
| [O-]I(=O)=O.[K+] |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature