Learn More
Potassium iodate, 98%, extra pure
CAS: 7758-05-6 | IKO3 | 214 g/mol
$131.42 - $131.42
Chemical Identifiers
| CAS | 7758-05-6 |
|---|---|
| Molecular Formula | IKO3 |
| Molecular Weight (g/mol) | 214 |
| MDL Number | MFCD00011406 |
| InChI Key | JLKDVMWYMMLWTI-UHFFFAOYSA-M |
| IUPAC Name | potassium iodate |
| SMILES | [K+].[O-][I](=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC196741000
|
Thermo Scientific Chemicals
196741000 |
100 g | Glass bottle |
Each for $131.42
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
| 7758-05-6 | |
| 214 | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| [K+].[O-][I](=O)=O |
| IKO3 | |
| MFCD00011406 | |
| potassium iodate |
Specifications
| 560°C | |
| White | |
| 100 g | |
| IKO3 | |
| MFCD00011406 | |
| 15, 7763 | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| potassium iodate | |
| 214 | |
| Extra Pure | |
| Glass bottle | |
| 3.9300g/mL |
| 7758-05-6,7732-18-5 | |
| Crystalline Powder | |
| 97.5% min. (Iodometry) | |
| KIO3 | |
| 1479 | |
| Solubility in water: soluble | |
| [K+].[O-][I](=O)=O | |
| 214 | |
| 98% | |
| 0.5% max. | |
| 3.93 | |
| Potassium iodate, 98% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 231-831-9
RTECSNumber : NN1350000
TSCA : TSCA
RUO – Research Use Only