missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution, 45% w/w (Certified), Fisher Chemical™

Catalog No. p-213795
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Poly Bottle
Poly Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SP236500 500 mL Poly Bottle
SP2364 4 L Poly Bottle
SP23620 20 L Poly Pail
SP236200 200 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SP236500 Supplier Thermo Fisher Scientific Supplier No. SP236 500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

pH 12.0
Color Colorless
Melting Point -28.8°C
Assay Percent Range 45 to 47 %
Density 1.510g/cm³
Quantity 500 mL
Formula Weight 56.11
Percent Purity 45 to 47%
Grade Certified
Concentration or Composition (by Analyte or Components) 45%
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 2.6mmHg at 20°C
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide Solution, 45% w/w
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive