missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution, 1N (In Methanol/Certified), Fisher Chemical

Catalog No. SP2201
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SP2201 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SP2201 Supplier Thermo Fisher Scientific Supplier No. FLSP2201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Titrated at 25°C to pH 8.2 (phenolphthalein endpoint) against standardized HCl

Chemical Identifiers

CAS 1310-58-3 , 67-56-1 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
Boiling Point 65.6°C
Melting Point -100°C
Concentration 0.995 to 1.005N
UN Number 2924
Density 0.8g/cm³
Quantity 1 L
Formula Weight 56.11
Grade Certified
Concentration or Composition (by Analyte or Components) 1N in Methanol
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide Solution, 1N, in methanol
Show More Show Less