missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution, 0.1N (N/10) (In Methanol/Certified), Fisher Chemical™

Catalog No. SP224P4
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SP224P4 4 L Poly Bottle
SP2241 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SP224P4 Supplier Thermo Fisher Scientific Supplier No. FLSP224P4
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1310-58-3 , 67-56-1 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

pH 13
Color Colorless
Boiling Point 66.1°C
Melting Point -100°C
Concentration 0.0995 to 0.1005N
UN Number 2924
Density 0.8g/cm³
Quantity 4 L
Formula Weight 56.11
Grade Certified
Concentration or Composition (by Analyte or Components) 0.1N (N/10)
Identification Pass Test
Packaging Poly Bottle
Traceability to NIST LOT 84L Potassium acid phthalate
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide Solution, 0.1N (N/10), in methanol
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation DOT Class 3, : Flammable Liquid