missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution, 0.1N (In Isopropanol/Certified), Fisher Chemical

Catalog No. ST110500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
HDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ST110500 500 mL HDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST110500 Supplier Thermo Fisher Scientific Supplier No. ST110 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use as a titration standard

Chemical Identifiers

CAS 1310-58-3 , 67-63-0
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Boiling Point 82.8°C
Melting Point -88.9°C
Concentration 0.0995 to 0.1005N
Density 0.8g/cm³
Quantity 500 mL
Grade Certified
Concentration or Composition (by Analyte or Components) 0.1N in Isopropanol
Identification Pass Test
Packaging HDPE Bottle
Traceability to NIST LOT 84L Potassium acid phthalate
Vapor Pressure 33mmHg at 20°C
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide Solution, 0.1N, in isopropanol
Show More Show Less