missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, Purified, Reagents

Catalog No. p-200021116
Change view
Click to view available options
Quantity:
500 g
3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337102 3 kg
01337101 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337102 Supplier Labchem Inc Supplier No. C22850003A5
Only null left

Potassium Hydroxide, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color White
Boiling Point 1320°C
Synonym Caustic Potash, Potash lye
Quantity 3 kg
Grade Pure
Packaging Poly Bottle
Specific Gravity 2.044
Physical Form Solid
Chemical Name or Material Potassium Hydroxide, Purified