missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, in Isopropanol, Certified, 0.500N ±0.005N (0.5M), LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 97.25, 2.75 |
| Linear Formula | KOH |
| Concentration | 0.500N ±0.005N (0.5M) |
| UN Number | UN2924 |
| Solubility Information | Soluble in water |
| Quantity | 500 mL |
| Formula Weight | 56.11 |
| Grade | Certified |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Highly flammable liquid and vapor. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. |
| Precautionary Statement | GHS P Statement Keep away from heat, sparks, open flames, hot surfaces. No smoking. Keep container tightly closed. Ground/bond container and receiving equipment. Use explosion-proof electrical, ventilating, lighting equipment. Use only non-sparking tools. Take precautionary measures against static discharge. Do not breathe mist, vapors, spray. Use only outdoors or in a well-ventilated area. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin thoroughly after handling. If swallowed: Rinse mounth. Do not induce vomiting. If on skin (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a poison center/doctor. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Immediately call a poison center/doctor. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish. Store locked up in a cool, well ventilated place. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |