missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide concentrate, For 1L standard solution, 1.0 M KOH (1.0N), Honeywell Fluka™

Catalog No. 60046689
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60046689 6 x 1 Ea.
6001256 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046689 Supplier Honeywell Chemicals Supplier No. 380736X1EA
Only null left

for 1L standard solution, 1.0 M KOH (1.0N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium;hydroxide
SMILES [OH-].[K+]

Specifications

Boiling Point 100°C
CAS Min % ≥25.0000%, ≥50.0000%
CAS Max % ≤50.0000%, ≤75.0000%
Linear Formula KOH
UN Number UN1814
Density 1.29 - 1.37 g/cm3
Quantity 6 x 1 Ea.
Formula Weight 56.11
Packaging Ampoule
Physical Form Concentrate
Chemical Name or Material Potassium hydroxide concentrate, For 1L standard solution, 1.0 M KOH (1.0N)
Show More Show Less
Hazard Statement P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Potassium hydroxide, solution
Recommended Storage Room Temp