missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, Certified, 2.00N ±0.02N (2.0M), LabChem™

Catalog No. LC193602
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
4 L
Catalog No. Quantity
LC193602 1 L
LC193604 4 L
2 options

Catalog No. LC193602

Supplier: LabChem LC193602

Only null left

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
CAS Max % 89.7, 10.3
Linear Formula KOH
Concentration 2.00N ±0.02N (2.0M)
UN Number UN1814
Density 1.09g/mL
Solubility Information Soluble in water
Quantity 1 L
Formula Weight 56.11
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.09g/mL
Traceability to NIST Traceable to NIST
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature