missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, Certified, 10.0N ±0.1N (10M), LabChem™

Catalog No. p-5389004
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC193701 500 mL
LC193702 1 L
LC193704 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC193701 Supplier LabChem Supplier No. LC193701
Only null left

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
CAS Max % 59.69, 40.31
Linear Formula KOH
Concentration 10.0N ±0.1N (10M)
UN Number UN1814
Density 1.39g/mL
Solubility Information Soluble in water
Quantity 500 mL
Formula Weight 56.11
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.39g/mL
Traceability to NIST Traceable to NIST
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container in accordance with local, state, and federal regulations.
Recommended Storage Room Temperature