missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide, ca. 50% solution in water

Catalog No. AC396060025
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC396060025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC396060025 Supplier Thermo Scientific Chemicals Supplier No. 396060025

CAS: 1310-58-3 | HKO | 56.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Linear Formula KOH
Density 1.3260g/mL
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and glycerol
Quantity 2.5 kg
Formula Weight 56.11
Packaging Plastic bottle
Specific Gravity 1.326
Chemical Name or Material Potassium hydroxide

RUO – Research Use Only