missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, ACS Reagent, Reagents

Catalog No. p-200020940
Change view
Click to view available options
Quantity:
100 mL
500 g
1 kg
2.5 kg
3 kg
10 kg
12 kg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337842 100 mL
01337841 10 kg
01337840 12 kg
01337839 1 kg
01337838 2.5 kg
01337837 3 kg
01337836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01337842 Supplier Labchem Inc Supplier No. C1285000100A6
Only null left

Potassium Hydroxide, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym Caustic Potash, Potash Lye
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color White
Boiling Point 1320°C
Quantity 100 mL
Grade ACS
Packaging Poly Bottle
Specific Gravity 2.044
Physical Form Solid
Chemical Name or Material Potassium Hydroxide, ACS Reagent