missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, ACS Reagent, Reagents
Potassium Hydroxide, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C12850001KG
Specifications
| Potassium Hydroxide, ACS Reagent | |
| 1320°C | |
| HKO | |
| Caustic Potash, Potash Lye | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| potassium hydroxide | |
| ACS | |
| 2.044 |
| White | |
| 1310-58-3,7732-18-5 | |
| MFCD00003553 | |
| 1 kg | |
| [OH-].[K+] | |
| 56.11 | |
| Poly Bottle | |
| Solid |
Chemical Identifiers
| 1310-58-3 | |
| 56.11 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| potassium hydroxide |
| HKO | |
| MFCD00003553 | |
| Caustic Potash, Potash Lye | |
| [OH-].[K+] |