missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, ACS Reagent, Reagents

Catalog No. 01337842
Change view
Click to view available options
Quantity:
100 mL
500 g
1 kg
2.5 kg
3 kg
10 kg
12 kg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337842 100 mL
01337841 10 kg
01337840 12 kg
01337839 1 kg
01337838 2.5 kg
01337837 3 kg
01337836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01337842 Supplier Labchem Inc Supplier No. C1285000100A6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Hydroxide, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym Caustic Potash, Potash Lye
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color White
Boiling Point 1320°C
Quantity 100 mL
Percent Purity 85%
Grade ACS
Packaging Poly Bottle
Specific Gravity 2.044
Physical Form Solid
Chemical Name or Material Potassium Hydroxide, ACS Reagent