missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, ACS Reagent Grade, Ricca Chemical

Catalog No. p-3252170
Change view
Click to view available options
Quantity:
500 g
25 kg
Packaging:
Fiber Drum
White Poly Wide Mouth Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCP0500500 500 g White Poly Wide Mouth Bottle
RDCP050025F 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RDCP0500500 Supplier Ricca Chemical Company Supplier No. RDCP0500500B1
Only null left

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color White
CAS Min % 98.0
CAS Max % 100.0
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Quantity 500 g
Percent Purity ≥85%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Physical Form Deliquescent Pellets
Chemical Name or Material Potassium Hydroxide
ShelfLife 120 months