Learn More
Potassium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.1N, Thermo Scientific Chemicals
$737.90 - $737.90
Chemical Identifiers
| CAS | 1310-58-3 |
|---|---|
| Molecular Formula | HKO |
| Molecular Weight (g/mol) | 56.11 |
| MDL Number | MFCD00003553 |
| InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
| Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
| PubChem CID | 14797 |
| ChEBI | CHEBI:32035 |
| SMILES | [OH-].[K+] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1310-58-3 | |
| 56.11 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| 14797 | |
| [OH-].[K+] |
| HKO | |
| MFCD00003553 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| CHEBI:32035 |
Specifications
| Colorless | |
| HKO | |
| UN1814 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| potassium hydroxide | |
| 14797 | |
| 56.11 | |
| Liquid |
| 1310-58-3 | |
| MFCD00003553 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| [OH-].[K+] | |
| 56.11 | |
| CHEBI:32035 | |
| 6 U | |
| Potassium hydroxide Acculute Standard Volumetric Solution, Final Concentration 0.1N |
Safety and Handling
GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POTASSIUM HYDROXIDE, SOLUTION
EINECSNumber : 215-181-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only