missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 8 Normal, Reagents

Catalog No. 01336176
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336176 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336176 Supplier Labchem Inc Supplier No. C84012NE1A
Only null left

Potassium Hydroxide, 8 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 1310-58-3
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Caustic Potash, Potash Lye
IUPAC Name water
SMILES O

Specifications

pH 14
Color Colorless
Boiling Point 135°C
Quantity 1 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.3229
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 8 Normal