missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 45% w/w, Reagent, Reagents

Catalog No. 01336446
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336446 1 L
01336445 4 L
01336444 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336446 Supplier Labchem Inc Supplier No. C72850001A
Only null left

Potassium Hydroxide, 45% w/w, Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1310-58-3
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Caustic Potash, Potash Lye
IUPAC Name water
SMILES O

Specifications

pH 14
Color Colorless
Boiling Point 130°C
Quantity 1 L
Percent Purity Min. 45%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.46
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 45% w/w, Reagent