missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, 45% (w/w) Aqueous Solution, Spectrum™ Chemical
Specifications
Specifications
| Concentration | 45% |
| Quantity | 4 L |
| Percent Purity | 45% |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Chemical Name or Material | Potassium Hydroxide Solution |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.