missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 45% (w/w) Aqueous Solution, Spectrum™ Chemical

Catalog No. 18612712
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Poly Bottle
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612712 500 mL Poly Bottle
18612713 4 L Poly Bottle
18612714 20 L Poly Drum
18612715 200 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612712 Supplier Spectrum Chemical Mfg Cor Supplier No. P1320500MLPL
Only null left

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5

Specifications

Concentration 45%
Quantity 500 mL
Percent Purity 45%
Grade Reagent
Packaging Poly Bottle
Chemical Name or Material Potassium Hydroxide Solution

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.