missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 45% (w/w), Analytical Reagent Grade, Ricca Chemical

Catalog No. R619000055E
Change view
Click to view available options
Quantity:
500 mL
1 L
20 L
55 gal.
Packaging:
Cubitainer
Natural Poly Bottle
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R619000055E 55 gal. Poly Drum
619016 500 mL Natural Poly Bottle
619032 1 L Natural Poly Bottle
61905 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R619000055E Supplier Ricca Chemical Company Supplier No. R619000055E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

pH >12
Color Colorless
Assay Percent Range 40% (w/v)
Solubility Information Solubility in Water: Infinite
Quantity 55 gal.
Percent Purity min 45% w/w
Grade Laboratory
Odor Odorless
Packaging Poly Drum
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide Solution
Show More Show Less
Recommended Storage Corrosive