missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 45% (w/w), Analytical Reagent Grade, Ricca Chemical

Catalog No. 619016
Change view
Click to view available options
Quantity:
500 mL
1 L
55 gal.
Packaging:
Natural Poly Bottle
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
619016 500 mL Natural Poly Bottle
619032 1 L Natural Poly Bottle
61905 - -
R619000055E 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 619016 Supplier Ricca Chemical Company Supplier No. 619016

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
CAS Min % 98.0
CAS Max % 100.0
Concentration or Composition Notes 45% (w/w)
Quantity 500 mL
Percent Purity Min. 45.0%
Grade Analytical
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide
Show More Show Less