missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 40% (w/v), Ricca Chemical

Catalog No. p-4665630
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R6156000500 500 mL Natural Poly Bottle
R61560001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R6156000500 Supplier Ricca Chemical Company Supplier No. R6156000500A
Only null left

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

pH >12
Color Colorless
Assay Percent Range 40% (w/v)
Solubility Information Solubility in Water: Infinite
Quantity 500 mL
Percent Purity 39.6 to 40.4% w/v
Grade Laboratory
Odor Odorless
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide Solution
Show More Show Less
Recommended Storage Corrosive