missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 4.00 N, Ricca Chemical

Supplier:  Ricca Chemical Company R6252000500A

Catalog No. R6252000500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Potassium Hydroxide
65.0,32.68
HKO
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium;hydroxide
14797
Natural Poly Bottle
4.00Normal
1310-58-3 , 7732-18-5
68.0,34.01
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
[OH-].[K+]
56.105
CHEBI:32035
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions